N,N-Diisopropylethylendiamin 97%

Synonyme: 2-Aminoethyldiisopropylamine,N,N-Bis(isopropyl)ethane-1,2-diamine

BD142120-5G BD142120-25G
BLDPBD142120-5GEA 7 EUR
BLDPBD142120-5G BLDPBD142120-25G
N,N-Diisopropylethylendiamin 97%
N,N-Diisopropylethylendiamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₂₀N₂
Molecular Weight: 144,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015015
CAS-Nummer: 121-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR