N,N-Diethylbutan-1-amine 95%

BLDPBD142052-5GEA 37 EUR
BLDPBD142052-5G BLDPBD142052-25G BLDPBD142052-1G
N,N-Diethylbutan-1-amine 95%
N,N-Diethylbutan-1-amine

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₉N
Molecular Weight: 129,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048759
CAS-Nummer: 4444-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR