N,N-Diethylacetoacetamid 98%

BLDPBD142042-5GEA 11 EUR
BLDPBD142042-5G BLDPBD142042-25G
N,N-Diethylacetoacetamid 98%
N,N-Diethylacetoacetamid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₅NO₂
Molecular Weight: 157,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026728
CAS-Nummer: 2235-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR