N,N-Dibenzylformamide 98%

BLDPBD141971-10GEA 35 EUR
BLDPBD141971-10G BLDPBD141971-0.025 BLDPBD141971-100G BLDPBD141971-25G BLDPBD141971-5G
N,N-Dibenzylformamide 98%
N,N-Dibenzylformamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₅NO
Molecular Weight: 225,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858481
CAS-Nummer: 5464-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR