N,N-Dibenzyl-2-aminoethanol 98%

Synonyme: N-(2-Hydroxyethyl)dibenzylamine,N,N-Dibenzylethanolamine

BD141962-500G BD141962-100G BD141962-5G BD141962-25G
BLDPBD141962-500GEA 645 EUR
BLDPBD141962-500G BLDPBD141962-100G BLDPBD141962-5G BLDPBD141962-25G
N,N-Dibenzyl-2-aminoethanol 98%
N,N-Dibenzyl-2-aminoethanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₉NO
Molecular Weight: 241,33 g/mol
Lagertemperatur: Ambient
MDL: MFCD00020574
CAS-Nummer: 101-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR