Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate 98%

Synonyme: N,N-Di-(beta-carboethoxyethyl)methylamine

BD141933-250MG
BLDPBD141933-250MGEA 6 EUR
BLDPBD141933-250MG
Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate 98%
Ethyl-3-[(3-ethoxy-3-oxopropyl)(methyl)amino]propanoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₂₁NO₄
Molecular Weight: 231,29 g/mol
Lagertemperatur: Ambient
MDL: MFCD00026916
CAS-Nummer: 6315-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR