N,N'-Bis(salicylidene)-1,2-propanediamine 97%

Synonyme: N,N'-Bis(2-hydroxybenzylidene)-1,2-propanediamine,N,N'-Disalicylal-1,2-propanediamine

BD141897-5G BD141897-10G BD141897-1G BD141897-100G BD141897-25G
BLDPBD141897-5GEA 19 EUR
BLDPBD141897-5G BLDPBD141897-10G BLDPBD141897-1G BLDPBD141897-100G BLDPBD141897-25G
N,N'-Bis(salicylidene)-1,2-propanediamine 97%
N,N'-Bis(salicylidene)-1,2-propanediamine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₇H₁₈N₂O₂
Molecular Weight: 282,34 g/mol
Lagertemperatur: Ambient
MDL: MFCD00020104
CAS-Nummer: 94-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR