2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%

BD141820-250MG BD141820-1G BD141820-100MG
BLDPBD141820-250MGEA 187 EUR
BLDPBD141820-250MG BLDPBD141820-1G BLDPBD141820-100MG
2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone 95%
2,9-Di(tridecan-7-yl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

Storage condition: Sealed in dry, 2-8°C

Formel: C₅₀H₆₂N₂O₄
Molecular Weight: 755,03 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08702668
CAS-Nummer: 110590-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR