(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine 98%

Synonyme: 2-Aminomethy-4-(4-fluorobenzyl)morpholine

BD14178-5G BD14178-10G BD14178-100G BD14178-25G BD14178-1G
BLDPBD14178-5GEA 37 EUR
BLDPBD14178-5G BLDPBD14178-10G BLDPBD14178-100G BLDPBD14178-25G BLDPBD14178-1G
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine 98%
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₂H₁₇FN₂O
Molecular Weight: 224,28 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD06658145
CAS-Nummer: 112914-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR