N,N,2-Trimethylquinolin-6-amine 97%

BLDPBD141741-100MGEA 23 EUR
BLDPBD141741-100MG BLDPBD141741-250MG BLDPBD141741-5G BLDPBD141741-1G
N,N,2-Trimethylquinolin-6-amine 97%
N,N,2-Trimethylquinolin-6-amine

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₄N₂
Molecular Weight: 186,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089173
CAS-Nummer: 92-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR