2-Isopropyl-N,2,3-trimethylbutyramide 97%

Synonyme: N,2,3-Trimethyl-2-isopropylbutamide,2-Isopropyl-N,2,3-trimethylbutanamide

BD141724-500G BD141724-10G BD141724-1000G BD141724-5G BD141724-25G BD141724-100G
BLDPBD141724-500GEA 263 EUR
BLDPBD141724-500G BLDPBD141724-10G BLDPBD141724-1000G BLDPBD141724-5G BLDPBD141724-25G BLDPBD141724-100G
2-Isopropyl-N,2,3-trimethylbutyramide 97%
2-Isopropyl-N,2,3-trimethylbutyramide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₁NO
Molecular Weight: 171,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00130070
CAS-Nummer: 51115-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR