N-(tert-Butoxycarbonyl)-L-serine-β-lactone 97%

Synonyme: (S)-ɑ-tert-Butoxycarbonylamino-β-propiolactone,tert-Butyl (S)-(2-oxooxetan-3-yl)carbamate,N-Boc-L-serineβ-Lactone,(S)-ɑ-Boc-amino-β-propiolactone,(S)-3-(tert-Butoxycarbonylamino)-2-oxetanone

BD141700-250MG
BLDPBD141700-250MGEA 25 EUR
BLDPBD141700-250MG
N-(tert-Butoxycarbonyl)-L-serine-β-lactone 97%
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₁₃NO₄
Molecular Weight: 187,20 g/mol
Lagertemperatur: Freezer
MDL: MFCD01318414
CAS-Nummer: 98541-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR