(3,4-Dimethoxyphenyl)essigsäureethylester 98%

Synonyme: Ethyl-(3,4-dimethoxyphenyl)acetat

BD14164-25G BD14164-10G BD14164-5G
BLDPBD14164-25GEA 51 EUR
BLDPBD14164-25G BLDPBD14164-10G BLDPBD14164-5G
(3,4-Dimethoxyphenyl)essigsäureethylester 98%
(3,4-Dimethoxyphenyl)essigsäureethylester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₄
Molecular Weight: 224,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00017272
CAS-Nummer: 18066-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR