1,3-Adamantandicarbonsäure 97%

Synonyme: 1,3-Dicarboxyadamantane,(1s,3s,5r,7r)-Adamantane-1,3-dicarboxylic acid

BD14161-100G BD14161-500G BD14161-5G BD14161-10G BD14161-25G BD14161-1G
BLDPBD14161-100GEA 209 EUR
BLDPBD14161-100G BLDPBD14161-500G BLDPBD14161-5G BLDPBD14161-10G BLDPBD14161-25G BLDPBD14161-1G
1,3-Adamantandicarbonsäure 97%
1,3-Adamantanedicarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₄
Molecular Weight: 224,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00167790
CAS-Nummer: 39269-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR