4,4'-Di-tert-butylbenzophenon 97%

Synonyme: Bis(4-(tert-butyl)phenyl)methanone

BD1416-1G BD1416-5G BD1416-250MG
BLDPBD1416-1GEA 80 EUR
BLDPBD1416-1G BLDPBD1416-5G BLDPBD1416-250MG
4,4'-Di-tert-butylbenzophenon 97%
4,4'-Di-tert-butylbenzophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₂₆O
Molecular Weight: 294,4306 g/mol
Lagertemperatur: Ambient
MDL: MFCD01319937
CAS-Nummer: 15796-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR