3-Acetamido-6-chloropyridazine 98%

BLDPBD141553-250MGEA 18 EUR
BLDPBD141553-250MG BLDPBD141553-1G
3-Acetamido-6-chloropyridazine 98%
3-Acetamido-6-chloropyridazine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₆ClN₃O
Molecular Weight: 171,584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835313
CAS-Nummer: 14959-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR