N-(4-Methoxybenzyliden)anilin 98+%

BD141430-5G BD141430-25G BD141430-1G
BLDPBD141430-5GEA 42 EUR
BLDPBD141430-5G BLDPBD141430-25G BLDPBD141430-1G
N-(4-Methoxybenzyliden)anilin 98+%
N-(4-Methoxybenzyliden)anilin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₁₃NO
Molecular Weight: 211,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00025836
CAS-Nummer: 836-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR