(S)-1-(4-Fluorobenzoyl)pyrrolidine-2-carboxylic acid 98%

BD141387-1G BD141387-250MG BD141387-5G
BLDPBD141387-1GEA 61 EUR
BLDPBD141387-1G BLDPBD141387-250MG BLDPBD141387-5G
(S)-1-(4-Fluorobenzoyl)pyrrolidine-2-carboxylic acid 98%
(S)-1-(4-Fluorobenzoyl)pyrrolidine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₂FNO₃
Molecular Weight: 237,23 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09757450
CAS-Nummer: 62522-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR