N-(4-Chlorophenyl)formamide 98%

BLDPBD141357-1GEA 45 EUR
BLDPBD141357-1G BLDPBD141357-0.025 BLDPBD141357-5G BLDPBD141357-25G
N-(4-Chlorophenyl)formamide 98%
N-(4-Chlorophenyl)formamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₆ClNO
Molecular Weight: 155,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021020
CAS-Nummer: 2617-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR