N-(4-Bromo-2,6-dichlorophenyl)acetamide 98%

BD141295-1G BD141295-100MG BD141295-250MG BD141295-5G
BLDPBD141295-1GEA 62 EUR
BLDPBD141295-1G BLDPBD141295-100MG BLDPBD141295-250MG BLDPBD141295-5G
N-(4-Bromo-2,6-dichlorophenyl)acetamide 98%
N-(4-Bromo-2,6-dichlorophenyl)acetamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆BrCl₂NO
Molecular Weight: 282,95 g/mol
Lagertemperatur: Ambient
MDL: MFCD08703152
CAS-Nummer: 13953-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR