1-Benzyl-1,4-diazepan-5-one 98%

Synonyme: 1-Benzylhomopiperazin-5-one

BD14120-250MG BD14120-5G BD14120-1G
BLDPBD14120-250MGEA 22 EUR
BLDPBD14120-250MG BLDPBD14120-5G BLDPBD14120-1G
1-Benzyl-1,4-diazepan-5-one 98%
1-Benzyl-1,4-diazepan-5-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆N₂O
Molecular Weight: 204,2700 g/mol
Lagertemperatur: Ambient
MDL: MFCD00225259
CAS-Nummer: 55186-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR