N-(3-Hydroxypropyl)phthalimid 95%

Synonyme: N-(3-Hydroxypropyl)-1,2-benzoldicarbonsäureimidN-(3-Hydroxypropyl)phthalsäureimid

BLDPBD141197-100GEA 167 EUR
BLDPBD141197-100G BLDPBD141197-5G BLDPBD141197-10G BLDPBD141197-25G BLDPBD141197-1G
N-(3-Hydroxypropyl)phthalimid 95%
N-(3-Hydroxypropyl)phthalimid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023097
CAS-Nummer: 883-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR