N-(3-Chloroquinoxalin-2-yl)-4-methylbenzenesulphonamide 95%

BD141170-1G
BLDPBD141170-1GEA 11 EUR
BLDPBD141170-1G
N-(3-Chloroquinoxalin-2-yl)-4-methylbenzenesulphonamide 95%
N-(3-Chloroquinoxalin-2-yl)-4-methylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₂ClN₃O₂S
Molecular Weight: 333,79 g/mol
Lagertemperatur: Ambient
MDL: MFCD00501967
CAS-Nummer: 4029-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR