N-(2-Pyridylmethyl)glycine ethyl ester 95%

BLDPBD141074-5GEA 113 EUR
BLDPBD141074-5G BLDPBD141074-1G
N-(2-Pyridylmethyl)glycine ethyl ester 95%
N-(2-Pyridylmethyl)glycine ethyl ester

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194,23 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00191573
CAS-Nummer: 62402-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR