N-[2-(Dimethylamino)ethyl]-N-methylformamide 98%

Synonyme: N-(2-(Dimethylamino)ethyl)-N-methylformamide,N-Formyl-N,N',N'-trimethylethylenediamine

BD140962-5G BD140962-1G BD140962-100G BD140962-25G
BLDPBD140962-5GEA 37 EUR
BLDPBD140962-5G BLDPBD140962-1G BLDPBD140962-100G BLDPBD140962-25G
N-[2-(Dimethylamino)ethyl]-N-methylformamide 98%
N-[2-(Dimethylamino)ethyl]-N-methylformamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₁₄N₂O
Molecular Weight: 130,19 g/mol
Lagertemperatur: Ambient
MDL: MFCD04039895
CAS-Nummer: 105669-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR