2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione 95%

BLDPBD140921-1GEA 15 EUR
BLDPBD140921-1G BLDPBD140921-250MG BLDPBD140921-25G BLDPBD140921-5G
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione 95%
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,63 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00129356
CAS-Nummer: 6270-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR