N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride 95%

BLDPBD140920-50MGEA 188 EUR
BLDPBD140920-50MG BLDPBD140920-100MG BLDPBD140920-25MG BLDPBD140920-250MG
N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride 95%
N-(2-Bromobenzyl)-2-chloro-N-ethylethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₆BrCl₂N
Molecular Weight: 313,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043241
CAS-Nummer: 40616-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR