1-Benzyl-L-glutamate 97%

Synonyme: L-Glutamic Acid 1-Benzyl Ester,H-Glu-OBzl,(S)-4-Amino-5-(benzyloxy)-5-oxopentanoic acid

BD14089-5G BD14089-100G BD14089-1G BD14089-25G
BLDPBD14089-5GEA 64 EUR
BLDPBD14089-5G BLDPBD14089-100G BLDPBD14089-1G BLDPBD14089-25G
1-Benzyl-L-glutamate 97%
1-Benzyl-L-glutamate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₅NO₄
Molecular Weight: 237,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00037312
CAS-Nummer: 13030-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR