2',4'-Dichlor-5'-hydroxyacetanilid 95%

Synonyme: N-(2,4-Dichloro-5-hydroxyphenyl)acetamide

BD140759-5G BD140759-1G BD140759-250MG
BLDPBD140759-5GEA 103 EUR
BLDPBD140759-5G BLDPBD140759-1G BLDPBD140759-250MG
2',4'-Dichlor-5'-hydroxyacetanilid 95%
2',4'-Dichlor-5'-hydroxyacetanilid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇Cl₂NO₂
Molecular Weight: 220,0527 g/mol
Lagertemperatur: Ambient
MDL: MFCD00186236
CAS-Nummer: 67669-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR