Butyl myristate 98%

BLDPBD140589-5GEA 13 EUR
BLDPBD140589-5G BLDPBD140589-100G BLDPBD140589-25G
Butyl myristate 98%
Butyl myristate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₈H₃₆O₂
Molecular Weight: 284,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015077
CAS-Nummer: 110-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR