4-(4-Methylpent-3-en-1-yl)cyclohex-3-ene-1-carbaldehyde Gemisch aus Isomeren 98% +

BD140578-25G
BLDPBD140578-25GEA 9 EUR
BLDPBD140578-25G
4-(4-Methylpent-3-en-1-yl)cyclohex-3-ene-1-carbaldehyde Gemisch aus Isomeren 98% +
4-(4-Methylpent-3-en-1-yl)cyclohex-3-ene-1-carbaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₂₀O
Molecular Weight: 192,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00169827
CAS-Nummer: 37677-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR