2-(1-Piperidinyl)-5-(trifluoromethyl)aniline 95%

Synonyme: 2-(Piperidin-1-yl)-5-(trifluoromethyl)aniline

BD14052-250MG BD14052-5G BD14052-10G BD14052-1G
BLDPBD14052-250MGEA 14 EUR
BLDPBD14052-250MG BLDPBD14052-5G BLDPBD14052-10G BLDPBD14052-1G
2-(1-Piperidinyl)-5-(trifluoromethyl)aniline 95%
2-(1-Piperidinyl)-5-(trifluoromethyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₅F₃N₂
Molecular Weight: 244,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00042161
CAS-Nummer: 1496-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR