Clofibrat 98%

Synonyme: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

BD14049-5G BD14049-25G BD14049-100G BD14049-1G
BLDPBD14049-5GEA 11 EUR
BLDPBD14049-5G BLDPBD14049-25G BLDPBD14049-100G BLDPBD14049-1G
Clofibrat 98%
Clofibrate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₅ClO₃
Molecular Weight: 242,70 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00000615
CAS-Nummer: 637-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR