4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride 97%

BLDPBD14045-250MGEA 40 EUR
BLDPBD14045-250MG BLDPBD14045-1G
4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride 97%
4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₅ClF₃N
Molecular Weight: 265,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02178913
CAS-Nummer: 6652-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR