4-Formylbenzeneboronic acid, neopentyl glycol ester 98%

Synonyme: 5,5-Dimethyl-2-(4-formylphenyl)-1,3,2-dioxaborinane,4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzaldehyde

BD14039-1G BD14039-250MG BD14039-5G
BLDPBD14039-1GEA 18 EUR
BLDPBD14039-1G BLDPBD14039-250MG BLDPBD14039-5G
4-Formylbenzeneboronic acid, neopentyl glycol ester 98%
4-Formylbenzeneboronic acid, neopentyl glycol ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₅BO₃
Molecular Weight: 218,057 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD06796332
CAS-Nummer: 128376-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR