Moroxydine hydrochloride 98%

BLDPBD140384-1GEA 6 EUR
BLDPBD140384-1G BLDPBD140384-25G BLDPBD140384-10G BLDPBD140384-5G
Moroxydine hydrochloride 98%
Moroxydine hydrochloride
Formel: C₆H₁₃N₅O
Molecular Weight: 207,66 g/mol
MDL: MFCD00012798
CAS-Nummer: 3160-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR