3-(N-Morpholino)propanesulphonic acid, hemisodium salt 97%

Synonyme: 3-Morpholinopropane-1-sulphonic acid, sodium salt(2:1),MOPS hemisodium salt

BD140360-25G
BLDPBD140360-25GEA 10 EUR
BLDPBD140360-25G
3-(N-Morpholino)propanesulphonic acid, hemisodium salt 97%
3-(N-Morpholino)propanesulphonic acid, hemisodium salt

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₃₀N₂NaO₈S₂
Molecular Weight: 441,5200 g/mol
Lagertemperatur: Ambient
MDL: MFCD00079461
CAS-Nummer: 117961-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR