tert-Butyl hydrogen succinate 97%

Synonyme: Succinic acid mono-tert-butyl ester,4-(tert-Butoxy)-4-oxobutanoic acid,tert-Butyl Hydrogen Succinate,Mono-tert-butyl Succinate

BD140346-500G BD140346-100G BD140346-25G BD140346-1G BD140346-10G BD140346-5G
BLDPBD140346-500GEA 801 EUR
BLDPBD140346-500G BLDPBD140346-100G BLDPBD140346-25G BLDPBD140346-1G BLDPBD140346-10G BLDPBD140346-5G
tert-Butyl hydrogen succinate 97%
tert-Butyl hydrogen succinate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₄O₄
Molecular Weight: 174,196 g/mol
Lagertemperatur: Ambient
MDL: MFCD00273441
CAS-Nummer: 15026-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR