Kaliumdihydrogenphosphat 99%

Synonyme: Kaliumphosphat monobasisch

BLDPBD140341-100GEA 6 EUR
BLDPBD140341-100G BLDPBD140341-1000G BLDPBD140341-500G
Kaliumdihydrogenphosphat 99%
Kaliumdihydrogenphosphat

Storage condition: Sealed in dry, Room Temperature

Formel: KH₂PO₄
Molecular Weight: 136,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR