1-Monodecanoin 95%

Synonyme: 2,3-Dihydroxypropyl decanoate

BD140299-5G BD140299-250MG BD140299-10G BD140299-1G
BLDPBD140299-5GEA 61 EUR
BLDPBD140299-5G BLDPBD140299-250MG BLDPBD140299-10G BLDPBD140299-1G
1-Monodecanoin 95%
1-Monodecanoin

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₃H₂₆O₄
Molecular Weight: 246,3431 g/mol
Lagertemperatur: Freezer
MDL: MFCD00056656
CAS-Nummer: 2277-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR