Methyl-5-allyl-3-methoxysalicylate 98%

Synonyme: 5-Allyl-3-methoxysalicylic Acid Methyl Ester,Methyl 5-Allyl-2-hydroxy-3-methoxybenzoate,Methyl 5-allyl-2-hydroxy-3-methoxybenzoate,5-Allyl-2-hydroxy-3-methoxybenzoic Acid Methyl Ester

BD14028-25G BD14028-10G
BLDPBD14028-25GEA 39 EUR
BLDPBD14028-25G BLDPBD14028-10G
Methyl-5-allyl-3-methoxysalicylate 98%
Methyl-5-allyl-3-methoxysalicylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₄O₄
Molecular Weight: 222,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD04038819
CAS-Nummer: 85614-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR