Methylcarbamat 95%

Synonyme: Carbamidsäuremethylester

BLDPBD139370-25GEA 7 EUR
BLDPBD139370-0.025 BLDPBD139370-25G BLDPBD139370-500G BLDPBD139370-1000G BLDPBD139370-100G
Methylcarbamat 95%
Methylcarbamat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂H₅NO₂
Molecular Weight: 75,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007964
CAS-Nummer: 598-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR