Methyl-5-iodsalicylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD139168-25GEA 110 EUR
BLDPBD139168-25G BLDPBD139168-5G BLDPBD139168-1G BLDPBD139168-100G
Methyl-5-iodsalicylat 98%
Methyl-5-iodsalicylat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016462
CAS-Nummer: 4068-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR