Diphenylphosphinsäure 98%

BLDPBD13844-5GEA 6 EUR
BLDPBD13844-5G BLDPBD13844-25G BLDPBD13844-10G BLDPBD13844-500G BLDPBD13844-100G
Diphenylphosphinsäure 98%
Diphenylphosphinic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₁O₂P
Molecular Weight: 218,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002132
CAS-Nummer: 1707-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR