4-Phenoxyphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD13796-25GEA 41 EUR
BLDPBD13796-25G BLDPBD13796-5G BLDPBD13796-10G BLDPBD13796-500G BLDPBD13796-100G BLDPBD13796-1G
4-Phenoxyphenylboronsäure 98%
4-Phenoxyphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR