Lithiumhexafluorophosphat 97%

BLDPBD136468-5GEA 7 EUR
BLDPBD136468-5G BLDPBD136468-25G
Lithiumhexafluorophosphat 97%
Lithium hexafluorophosphate

Storage condition: Inert atmosphere, Room Temperature

Formel: F₆LiP
Molecular Weight: 151,91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011096
CAS-Nummer: 21324-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR