Isosorbid 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD135397-100GEA
11
EUR
BLDPBD135397-0.5G
BLDPBD135397-100G
BLDPBD135397-1000G
BLDPBD135397-25G
BLDPBD135397-500G
Isosorbid 98%
Isosorbid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₁₀O₄ Molecular Weight: 146,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064827 CAS-Nummer: 652-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...