4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol 95%

BLDPBD134872-25GEA 66 EUR
BLDPBD134872-25G BLDPBD134872-1G BLDPBD134872-10G BLDPBD134872-250MG BLDPBD134872-5G
4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol 95%
4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₃₃H₅₆N₄OS₂
Molecular Weight: 588,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134696
CAS-Nummer: 991-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR