5-Aminoimidazo[1,2-a]pyridin 98%

BLDPBD134388-5GEA 117 EUR
BLDPBD134388-5G BLDPBD134388-250MG BLDPBD134388-1G BLDPBD134388-25G BLDPBD134388-10G
5-Aminoimidazo[1,2-a]pyridin 98%
5-Aminoimidazo[1,2-a]pyridin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₇N₃
Molecular Weight: 133,1506 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863377
CAS-Nummer: 66358-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR