Hexaketocyclohexane octahydrate 99%
|
|
BLDPBD133248-25GEA
102
EUR
BLDPBD133248-25G
BLDPBD133248-100G
BLDPBD133248-1G
BLDPBD133248-10G
BLDPBD133248-5G
Hexaketocyclohexane octahydrate 99%
Hexaketocyclohexane octahydrate
Storage condition: Sealed in dry, Room Temperature
Formel:
C₆O₆·8H₂O Molecular Weight: 312,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149074 CAS-Nummer: 7255-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...